4-(Chloromethyl)-2-(4-methylphenyl)-1,3-thiazole

Supplier: Matrix Scientific

029506-500MG
101828-292EA 290.77 USD
101828-292
4-(Chloromethyl)-2-(4-methylphenyl)-1,3-thiazole
4-(Chloromethyl)-2-(4-methylphenyl)-1,3-thiazole

Hazard: Irritant

Formula: C₁₁H₁₀ClNS
MDL Number: MFCD03423391
CAS Number: 35199-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR