4-(Chloromethyl)-2-(2-chlorophenyl)-1,3-thiazole

Supplier: Matrix Scientific

029503-500MG
101828-286EA 217.35 USD
101828-286
4-(Chloromethyl)-2-(2-chlorophenyl)-1,3-thiazole
4-(Chloromethyl)-2-(2-chlorophenyl)-1,3-thiazole

Hazard: Irritant

Formula: C₁₀H₇Cl₂NS
MDL Number: MFCD03423388
CAS Number: 588676-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR