N-1,3-Benzothiazol-2-yl-2-chloroacetamide

Supplier: Matrix Scientific

029500-500MG
101828-280EA 144.9 USD
101828-280
N-1,3-Benzothiazol-2-yl-2-chloroacetamide
N-1,3-Benzothiazol-2-yl-2-chloroacetamide

Hazard: Irritant

Formula: C₉H₇ClN₂OS
Melting Pt: 160 °C
MDL Number: MFCD00022860
CAS Number: 3028-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR