2-Chloro-N-(4-chloro-3-nitrophenyl)acetamide

Supplier: Matrix Scientific

029497-500MG
101828-276EA 146.15 USD
101828-276
2-Chloro-N-(4-chloro-3-nitrophenyl)acetamide
2-Chloro-N-(4-chloro-3-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₈H₆Cl₂N₂O₃
MDL Number: MFCD00157377
CAS Number: 196935-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR