2-Chloro-N-(5-chloro-2-methylphenyl)acetamide

Supplier: Matrix Scientific

029496-500MG
101828-274EA 146.15 USD
101828-274
2-Chloro-N-(5-chloro-2-methylphenyl)acetamide
2-Chloro-N-(5-chloro-2-methylphenyl)acetamide

Hazard: Irritant

Formula: C₉H₉Cl₂NO
MDL Number: MFCD00018899
CAS Number: 85817-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR