2-Chloro-N-(2-chloro-5-nitrophenyl)acetamide

Supplier: Matrix Scientific

029495-500MG
101828-272EA 146.15 USD
101828-272
2-Chloro-N-(2-chloro-5-nitrophenyl)acetamide
2-Chloro-N-(2-chloro-5-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₈H₆Cl₂N₂O₃
MDL Number: MFCD01357758
CAS Number: 108086-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR