Dimethyl-5-[(chloroacetyl)amino]isophthalate

Supplier: Matrix Scientific

029492-500MG
101828-266EA 146.15 USD
101828-266
Dimethyl-5-[(chloroacetyl)amino]isophthalate
Dimethyl-5-[(chloroacetyl)amino]isophthalate

Hazard: Irritant

Formula: C₁₂H₁₂ClNO₅
MDL Number: MFCD00157396
CAS Number: 198488-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR