Dimethyl-2-[(chloroacetyl)amino]terephthalate

Supplier: Matrix Scientific

029491-500MG
101828-264EA 146.15 USD
101828-264
Dimethyl-2-[(chloroacetyl)amino]terephthalate
Dimethyl-2-[(chloroacetyl)amino]terephthalate

Hazard: Irritant

Formula: C₁₂H₁₂ClNO₅
MDL Number: MFCD02342053
CAS Number: 325763-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR