N-(4-Acetylphenyl)-2-chloroacetamide

Supplier: Matrix Scientific

029488-500MG
101828-258EA 146.15 USD
101828-258
N-(4-Acetylphenyl)-2-chloroacetamide
N-(4-Acetylphenyl)-2-chloroacetamide

Hazard: Irritant

Formula: C₁₀H₁₀ClNO₂
MDL Number: MFCD00460866
CAS Number: 38283-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR