N-(4-Bromo-2-chlorophenyl)-2-chloroacetamide
Supplier: Matrix Scientific
029486-500MG
101828-254EA
146.15
USD
101828-254
N-(4-Bromo-2-chlorophenyl)-2-chloroacetamide
N-(4-Bromo-2-chlorophenyl)-2-chloroacetamide
Hazard: Irritant
Formula:
C₈H₆BrCl₂NO |
MDL Number:
MFCD00157385 CAS Number: 195372-57-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...