2-Chloro-N-(2-ethoxyphenyl)acetamide

Supplier: Matrix Scientific

029485-500MG
101828-252EA 146.15 USD
101828-252
2-Chloro-N-(2-ethoxyphenyl)acetamide
2-Chloro-N-(2-ethoxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₂ClNO₂
MDL Number: MFCD00157389
CAS Number: 21118-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR