2-Chloro-N-[2-chloro-5-(trifluoromethyl)phenyl]acetamide

Supplier: Matrix Scientific

Synonyms: 2-Chloro-N-(2-chloro-5-(trifluoromethyl)phenyl)acetamide

029483-500MG
101828-248EA 146.15 USD
101828-248
2-Chloro-N-[2-chloro-5-(trifluoromethyl)phenyl]acetamide
2-Chloro-N-[2-chloro-5-(trifluoromethyl)phenyl]acetamide

Hazard: Irritant

Formula: C₉H₆Cl₂F₃NO
MW: 272.06 g/mol
MDL Number: MFCD00077472
CAS Number: 328-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR