2-Chloro-N-(4-chloro-2,5-dimethoxyphenyl)acetamide

Supplier: Matrix Scientific

029479-500MG
101828-240EA 146.15 USD
101828-240
2-Chloro-N-(4-chloro-2,5-dimethoxyphenyl)acetamide
2-Chloro-N-(4-chloro-2,5-dimethoxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₁Cl₂NO₃
MDL Number: MFCD01034345
CAS Number: 448250-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR