2-Chloro-N-(2,5-dimethoxyphenyl)acetamide

Supplier: Matrix Scientific

029478-500MG
101828-238EA 146.15 USD
101828-238
2-Chloro-N-(2,5-dimethoxyphenyl)acetamide
2-Chloro-N-(2,5-dimethoxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₂ClNO₃
MDL Number: MFCD00095383
CAS Number: 22158-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR