2-Chloro-N-(3-chloro-2-methylphenyl)acetamide
Supplier: Matrix Scientific
029473-500MG
101828-226EA
146.15
USD
101828-226
2-Chloro-N-(3-chloro-2-methylphenyl)acetamide
2-Chloro-N-(3-chloro-2-methylphenyl)acetamide
Hazard: Irritant
Formula:
C₉H₉Cl₂NO |
MDL Number:
MFCD00018897 CAS Number: 99585-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...