2-Chloro-N-(2,5-dimethylphenyl)acetamide

Supplier: Matrix Scientific

029466-500MG
101828-212EA 109.25 USD
101828-212
2-Chloro-N-(2,5-dimethylphenyl)acetamide
2-Chloro-N-(2,5-dimethylphenyl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₂ClNO
CAS Number: 5177-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR