Methyl-2-[(chloroacetyl)amino]-5-methyl-4-phenylthiophene-3-carboxylate

Supplier: Matrix Scientific

029451-500MG
101828-182EA 217.35 USD
101828-182
Methyl-2-[(chloroacetyl)amino]-5-methyl-4-phenylthiophene-3-carboxylate
Methyl-2-[(chloroacetyl)amino]-5-methyl-4-phenylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₄ClNO₃S
MDL Number: MFCD03423379
CAS Number: 610274-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR