Methyl-2-[(chloroacetyl)amino]-4-(4-methoxyphenyl)thiophene-3-carboxylate

Supplier: Matrix Scientific

029449-500MG
101828-178EA 290.77 USD
101828-178
Methyl-2-[(chloroacetyl)amino]-4-(4-methoxyphenyl)thiophene-3-carboxylate
Methyl-2-[(chloroacetyl)amino]-4-(4-methoxyphenyl)thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₄ClNO₄S
MDL Number: MFCD02853981
CAS Number: 667407-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR