Methyl-2-[(chloroacetyl)amino]-4-(2,5-dimethylphenyl)thiophene-3-carboxylate

Supplier: Matrix Scientific

029448-500MG
101828-176EA 290.77 USD
101828-176
Methyl-2-[(chloroacetyl)amino]-4-(2,5-dimethylphenyl)thiophene-3-carboxylate
Methyl-2-[(chloroacetyl)amino]-4-(2,5-dimethylphenyl)thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₆H₁₆ClNO₃S
MDL Number: MFCD02853975
CAS Number: 667407-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR