Methyl-2-[(chloroacetyl)amino]-4-(4-fluorophenyl)thiophene-3-carboxylate

Supplier: Matrix Scientific

029447-500MG
101828-174EA 290.77 USD
101828-174
Methyl-2-[(chloroacetyl)amino]-4-(4-fluorophenyl)thiophene-3-carboxylate
Methyl-2-[(chloroacetyl)amino]-4-(4-fluorophenyl)thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₁ClFNO₃S
MDL Number: MFCD02853996
CAS Number: 519016-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR