Methyl-2-[(chloroacetyl)amino]-4-(4-chlorophenyl)thiophene-3-carboxylate

Supplier: Matrix Scientific

029446-500MG
101828-172EA 217.35 USD
101828-172
Methyl-2-[(chloroacetyl)amino]-4-(4-chlorophenyl)thiophene-3-carboxylate
Methyl-2-[(chloroacetyl)amino]-4-(4-chlorophenyl)thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₁Cl₂NO₃S
MDL Number: MFCD02853977
CAS Number: 519016-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR