Ethyl-2-[(chloroacetyl)amino]-5-methylthiophene-3-carboxylate

Supplier: Matrix Scientific

029442-500MG
101828-164EA 217.35 USD
101828-164
Ethyl-2-[(chloroacetyl)amino]-5-methylthiophene-3-carboxylate
Ethyl-2-[(chloroacetyl)amino]-5-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₂ClNO₃S
MDL Number: MFCD03423378
CAS Number: 669747-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR