Methyl-2-[(chloroacetyl)amino]-5-ethylthiophene-3-carboxylate
Supplier: Matrix Scientific
029441-500MG
101828-162EA
290.77
USD
101828-162
Methyl-2-[(chloroacetyl)amino]-5-ethylthiophene-3-carboxylate
Methyl-2-[(chloroacetyl)amino]-5-ethylthiophene-3-carboxylate
Hazard: Irritant
Formula:
C₁₀H₁₂ClNO₃S |
MDL Number:
MFCD03423377 CAS Number: 515832-73-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...