Isopropyl 5-(aminocarbonyl)-2-[(chloroacetyl)amino]-4-methylthiophene-3-carboxylate

Supplier: Matrix Scientific

029439-500MG
101828-158EA 290.77 USD
101828-158
Isopropyl 5-(aminocarbonyl)-2-[(chloroacetyl)amino]-4-methylthiophene-3-carboxylate
Isopropyl 5-(aminocarbonyl)-2-[(chloroacetyl)amino]-4-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂O₄S
MDL Number: MFCD03423375
CAS Number: 669747-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR