Methyl-5-(aminocarbonyl)-2-[(chloroacetyl)amino]-4-methylthiophene-3-carboxylate

Supplier: Matrix Scientific

029436-500MG
101828-152EA 290.77 USD
101828-152
Methyl-5-(aminocarbonyl)-2-[(chloroacetyl)amino]-4-methylthiophene-3-carboxylate
Methyl-5-(aminocarbonyl)-2-[(chloroacetyl)amino]-4-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₁ClN₂O₄S
MDL Number: MFCD03423374
CAS Number: 669747-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR