Dimethyl-5-[(chloroacetyl)amino]-3-methylthiophene-2,4-dicarboxylate

Supplier: Matrix Scientific

029435-500MG
101828-150EA 217.35 USD
101828-150
Dimethyl-5-[(chloroacetyl)amino]-3-methylthiophene-2,4-dicarboxylate
Dimethyl-5-[(chloroacetyl)amino]-3-methylthiophene-2,4-dicarboxylate

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₅S
MDL Number: MFCD00610982
CAS Number: 314245-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR