Methyl-2-[(chloroacetyl)amino]-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate

Supplier: Matrix Scientific

029431-500MG
101828-142EA 290.77 USD
101828-142
Methyl-2-[(chloroacetyl)amino]-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
Methyl-2-[(chloroacetyl)amino]-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₆ClNO₃S
MDL Number: MFCD01357147
CAS Number: 438029-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR