2-Chloro-N-(3-cyano-5,6,7,8-tetrahydro-4H-cyclohepta[b]thien-2-yl)acetamide

Supplier: Matrix Scientific

029430-500MG
101828-140EA 290.77 USD
101828-140
2-Chloro-N-(3-cyano-5,6,7,8-tetrahydro-4H-cyclohepta[b]thien-2-yl)acetamide
2-Chloro-N-(3-cyano-5,6,7,8-tetrahydro-4H-cyclohepta[b]thien-2-yl)acetamide

Hazard: Irritant

Formula: C₁₂H₁₃ClN₂OS
MDL Number: MFCD02329393
CAS Number: 315684-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR