Isopropyl-2-[(chloroacetyl)amino]-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Supplier: Matrix Scientific

029429-500MG
101828-138EA 217.35 USD
101828-138
Isopropyl-2-[(chloroacetyl)amino]-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Isopropyl-2-[(chloroacetyl)amino]-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₈ClNO₃S
MDL Number: MFCD03385896
CAS Number: 560080-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR