Ethyl-2-[(chloroacetyl)amino]-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate

Supplier: Matrix Scientific

029425-500MG
101828-130EA 217.35 USD
101828-130
Ethyl-2-[(chloroacetyl)amino]-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
Ethyl-2-[(chloroacetyl)amino]-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₄ClNO₃S
MDL Number: MFCD00436345
CAS Number: 203385-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR