Methyl-2-[(chloroacetyl)amino]-4-ethyl-5-methylthiophene-3-carboxylate

Supplier: Matrix Scientific

029424-500MG
101828-128EA 290.77 USD
101828-128
Methyl-2-[(chloroacetyl)amino]-4-ethyl-5-methylthiophene-3-carboxylate
Methyl-2-[(chloroacetyl)amino]-4-ethyl-5-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₄ClNO₃S
MDL Number: MFCD03377626
CAS Number: 544428-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR