Ethyl-2-[(chloroacetyl)amino]-4,5-dimethylthiophene-3-carboxylate

Supplier: Matrix Scientific

029423-500MG
101828-126EA 290.77 USD
101828-126
Ethyl-2-[(chloroacetyl)amino]-4,5-dimethylthiophene-3-carboxylate
Ethyl-2-[(chloroacetyl)amino]-4,5-dimethylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₄ClNO₃S
MDL Number: MFCD00297823
CAS Number: 60442-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR