Methyl-2-[(chloroacetyl)amino]-4,5-dimethylthiophene-3-carboxylate

Supplier: Matrix Scientific

029422-500MG
101828-124EA 290.77 USD
101828-124
Methyl-2-[(chloroacetyl)amino]-4,5-dimethylthiophene-3-carboxylate
Methyl-2-[(chloroacetyl)amino]-4,5-dimethylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₂ClNO₃S
MDL Number: MFCD02342280
CAS Number: 448199-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR