2-[(Chloroacetyl)amino]-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide

Supplier: Matrix Scientific

029421-500MG
101828-122EA 290.77 USD
101828-122
2-[(Chloroacetyl)amino]-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
2-[(Chloroacetyl)amino]-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂O₂S
MDL Number: MFCD02341112
CAS Number: 436090-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR