Methyl-2-[(chloroacetyl)amino]-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Supplier: Matrix Scientific

029419-500MG
101828-118EA 217.35 USD
101828-118
Methyl-2-[(chloroacetyl)amino]-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Methyl-2-[(chloroacetyl)amino]-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₆ClNO₃S
MDL Number: MFCD01650650
CAS Number: 448226-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR