Diethyl-5-[(chloroacetyl)amino]-3-methylthiophene-2,4-dicarboxylate

Supplier: Matrix Scientific

029418-500MG
101828-116EA 217.35 USD
101828-116
Diethyl-5-[(chloroacetyl)amino]-3-methylthiophene-2,4-dicarboxylate
Diethyl-5-[(chloroacetyl)amino]-3-methylthiophene-2,4-dicarboxylate

Hazard: Irritant

Formula: C₁₃H₁₆ClNO₅S
MDL Number: MFCD00414318
CAS Number: 146381-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR