2-Chloro-N-[4-(4-chlorophenyl)-1,3-thiazol-2-yl]acetamide

Supplier: Matrix Scientific

029411-500MG
101828-102EA 290.77 USD
101828-102
2-Chloro-N-[4-(4-chlorophenyl)-1,3-thiazol-2-yl]acetamide
2-Chloro-N-[4-(4-chlorophenyl)-1,3-thiazol-2-yl]acetamide

Hazard: Irritant

Formula: C₁₁H₈Cl₂N₂OS
MDL Number: MFCD00124551
CAS Number: 6125-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR