2-(Chloromethyl)imidazo[1,2-a]pyridine

Supplier: Matrix Scientific

029408-500MG
101828-096EA 135.7 USD
101828-096
2-(Chloromethyl)imidazo[1,2-a]pyridine
2-(Chloromethyl)imidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₇ClN₂
MDL Number: MFCD00269177
CAS Number: 57892-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR