4-Cyclopropyl-1,3-thiazol-2-amine
Supplier: Matrix Scientific
Synonyms:
4-Cyclopropylthiazol-2-amine
029407-500MG
101828-094EA
56.92
USD
101828-094
4-Cyclopropyl-1,3-thiazol-2-amine
4-Cyclopropyl-1,3-thiazol-2-amine
Hazard: Irritant
Formula:
C₆H₈N₂S MW: 140.21 g/mol |
MDL Number:
MFCD01571854 CAS Number: 324579-90-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...