4-Isopropyl-1,3-thiazol-2-amine hydrobromide
Supplier: Matrix Scientific
Synonyms:
4-Isopropyl-1,3-thiazol-2-amine
029406-500MG
101828-092EA
238.46
USD
101828-092
4-Isopropyl-1,3-thiazol-2-amine hydrobromide
4-Isopropyl-1,3-thiazol-2-amine hydrobromide
Hazard: Irritant
Formula:
C₆H₁₁BrN₂S MW: 223.14 g/mol |
MDL Number:
MFCD21605754 CAS Number: 79932-20-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...