1-[(1,5-Dimethyl-1H-pyrazol-4-yl)methyl]piperazine

Supplier: Matrix Scientific

029405-500MG
101828-090EA 290.77 USD
101828-090
1-[(1,5-Dimethyl-1H-pyrazol-4-yl)methyl]piperazine
1-[(1,5-Dimethyl-1H-pyrazol-4-yl)methyl]piperazine

Hazard: Irritant

Formula: C₁₀H₁₈N₄
MDL Number: MFCD02254043
CAS Number: 1001757-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR