1-[(3-Chloro-4-methoxyphenyl)sulfonyl]piperazine

Supplier: Matrix Scientific

029404-500MG
101828-088EA 193.85 USD
101828-088
1-[(3-Chloro-4-methoxyphenyl)sulfonyl]piperazine
1-[(3-Chloro-4-methoxyphenyl)sulfonyl]piperazine

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O₃S
MDL Number: MFCD03444516
CAS Number: 524711-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR