1-[(1-Ethyl-3,5-dimethyl-1H-pyrazol-4-yl)methyl]piperazine

Supplier: Matrix Scientific

029402-500MG
101828-084EA 290.77 USD
101828-084
1-[(1-Ethyl-3,5-dimethyl-1H-pyrazol-4-yl)methyl]piperazine
1-[(1-Ethyl-3,5-dimethyl-1H-pyrazol-4-yl)methyl]piperazine

Hazard: Irritant

Formula: C₁₂H₂₂N₄
MDL Number: MFCD06739351
CAS Number: 1004195-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR