4-Amino-N-cyclopentylbenzamide

Supplier: Matrix Scientific

029399-500MG
101828-078EA 218.46 USD
101828-078
4-Amino-N-cyclopentylbenzamide
4-Amino-N-cyclopentylbenzamide

Hazard: Irritant

Formula: C₁₂H₁₆N₂O
MDL Number: MFCD04035385
CAS Number: 436095-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR