1-(4-Ethylbenzyl)piperazine

Supplier: Matrix Scientific

029393-500MG
101828-064EA 322 USD
101828-064
1-(4-Ethylbenzyl)piperazine
1-(4-Ethylbenzyl)piperazine

Hazard: Irritant

Formula: C₁₃H₂₀N₂
Boiling Pt: 307 °C
Flash Pt: 121.6 °C
MDL Number: MFCD03701709
CAS Number: 435341-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR