1-[(5-Methyl-2-furyl)methyl]piperazine

Supplier: Matrix Scientific

029392-500MG
101828-062EA 290.77 USD
101828-062
1-[(5-Methyl-2-furyl)methyl]piperazine
1-[(5-Methyl-2-furyl)methyl]piperazine

Hazard: Irritant

Formula: C₁₀H₁₆N₂O
MDL Number: MFCD07612572
CAS Number: 873433-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR