1-(4-Isobutoxy-3-methoxybenzyl)piperazine

Supplier: Matrix Scientific

029390-500MG
101828-056EA 290.77 USD
101828-056
1-(4-Isobutoxy-3-methoxybenzyl)piperazine
1-(4-Isobutoxy-3-methoxybenzyl)piperazine

Hazard: Irritant

Formula: C₁₆H₂₆N₂O₂
MDL Number: MFCD05189232
CAS Number: 523981-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR