1-(3-Isobutoxybenzyl)piperazine

Supplier: Matrix Scientific

029389-500MG
101828-054EA 290.77 USD
101828-054
1-(3-Isobutoxybenzyl)piperazine
1-(3-Isobutoxybenzyl)piperazine

Hazard: Irritant

Formula: C₁₅H₂₄N₂O
MDL Number: MFCD05189237
CAS Number: 523981-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR