1-(2-Isopropoxybenzyl)piperazine

Supplier: Matrix Scientific

029385-500MG
101828-042EA 290.77 USD
101828-042
1-(2-Isopropoxybenzyl)piperazine
1-(2-Isopropoxybenzyl)piperazine

Hazard: Irritant

Formula: C₁₄H₂₂N₂O
MDL Number: MFCD09434011
CAS Number: 906088-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR