1-(5-Chloro-2-methoxybenzyl)piperazine

Supplier: Matrix Scientific

029383-500MG
101828-038EA 217.35 USD
101828-038
1-(5-Chloro-2-methoxybenzyl)piperazine
1-(5-Chloro-2-methoxybenzyl)piperazine

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MDL Number: MFCD03030343
CAS Number: 523980-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR